Product Details
- SNP ID
-
rs200901437
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.1:180916618 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGGAGGCCCCAGGCTTCCCAGGC[C/T]GCCTGCCCCTGGACGTGAGTACTGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1614
PubMed Links
Gene Details
- Gene
- KIAA1614
- Gene Name
- KIAA1614
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020950.1 |
582 |
Missense Mutation |
CCG,CTG |
P172L |
NP_066001.1 |
View Full Product Details