Product Details
- SNP ID
-
rs200730801
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:151404876 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGGTCTCACTTTCACCCTCAAAGA[A/G]CTGGTGAAGACTTTCAGGCTCAATT
- Phenotype
-
MIM: 614787
MIM: 602177
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
POGZ
PubMed Links
Gene Details
- Gene
- POGZ
- Gene Name
- pogo transposable element with ZNF domain
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001194937.1 |
5033 |
Missense Mutation |
CTC,TTC |
L1378F |
NP_001181866.1 |
NM_001194938.1 |
5033 |
Missense Mutation |
CTC,TTC |
L1325F |
NP_001181867.1 |
NM_015100.3 |
5033 |
Missense Mutation |
CTC,TTC |
L1387F |
NP_055915.2 |
NM_145796.3 |
5033 |
Missense Mutation |
CTC,TTC |
L1292F |
NP_665739.3 |
NM_207171.2 |
5033 |
Missense Mutation |
CTC,TTC |
L1334F |
NP_997054.1 |
XM_005244999.2 |
5033 |
Missense Mutation |
CTC,TTC |
L1387F |
XP_005245056.1 |
XM_005245000.4 |
5033 |
Missense Mutation |
CTC,TTC |
L1387F |
XP_005245057.1 |
XM_005245001.2 |
5033 |
Missense Mutation |
CTC,TTC |
L1387F |
XP_005245058.1 |
XM_005245005.1 |
5033 |
Missense Mutation |
CTC,TTC |
L1334F |
XP_005245062.1 |
XM_005245006.4 |
5033 |
Missense Mutation |
CTC,TTC |
L1334F |
XP_005245063.1 |
XM_011509331.2 |
5033 |
Missense Mutation |
CTC,TTC |
L1268F |
XP_011507633.1 |
XM_017000744.1 |
5033 |
Missense Mutation |
CTC,TTC |
L1394F |
XP_016856233.1 |
XM_017000745.1 |
5033 |
Missense Mutation |
CTC,TTC |
L1378F |
XP_016856234.1 |
XM_017000746.1 |
5033 |
Missense Mutation |
CTC,TTC |
L1378F |
XP_016856235.1 |
XM_017000747.1 |
5033 |
Missense Mutation |
CTC,TTC |
L1338F |
XP_016856236.1 |
XM_017000748.1 |
5033 |
Missense Mutation |
CTC,TTC |
L1334F |
XP_016856237.1 |
XM_017000749.1 |
5033 |
Missense Mutation |
CTC,TTC |
L1334F |
XP_016856238.1 |
- Gene
- PSMB4
- Gene Name
- proteasome subunit beta 4
There are no transcripts associated with this gene.
View Full Product Details