Product Details

SNP ID
rs200531164
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:247256985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGATTCTCTGCTAGTGTGGCTCC[A/T]GGTTATTGTGGCCAATAGCTATGCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
VN1R5 PubMed Links

Gene Details

Gene
VN1R5
Gene Name
vomeronasal 1 receptor 5 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173858.1 914 Missense Mutation CAG,CTG Q305L NP_776257.1

View Full Product Details