Product Details
- SNP ID
-
rs200506377
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:42693500 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTCTGTCTTGCAGGAATGACACC[A/G]AGGAAGATGTATTTGTACACCAGGT
- Phenotype
-
MIM: 154030
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
YBX1
PubMed Links
Gene Details
- Gene
- YBX1
- Gene Name
- Y-box binding protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004559.4 |
572 |
Missense Mutation |
AAG,GAG |
K81E |
NP_004550.2 |
View Full Product Details