Product Details

SNP ID
rs200228790
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:101239068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACTATGATATCATCGTCCGGCA[C/T]TACAACTACACGGGAAAGCTGAATA
Phenotype
MIM: 601974
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928370 PubMed Links

Gene Details

Gene
LOC101928370
Gene Name
uncharacterized LOC101928370
There are no transcripts associated with this gene.

Gene
S1PR1
Gene Name
sphingosine-1-phosphate receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320730.1 490 Silent Mutation CAC,CAT H28H NP_001307659.1
NM_001400.4 490 Silent Mutation CAC,CAT H28H NP_001391.2

View Full Product Details