Product Details

SNP ID
rs200640848
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:155284620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGCATTGGCTATGGGCAGCAG[G/T]CACCTGTAGGCATGCCCGACGTCTG
Phenotype
MIM: 609973 MIM: 609712
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HCN3 PubMed Links

Gene Details

Gene
HCN3
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020897.2 328 Missense Mutation GCA,TCA A318S NP_065948.1
XM_011509816.2 328 Missense Mutation GCA,TCA A271S XP_011508118.1
XM_011509817.2 328 Missense Mutation GCA,TCA A264S XP_011508119.1
XM_011509818.2 328 Missense Mutation GCA,TCA A79S XP_011508120.1
XM_017001918.1 328 Missense Mutation GCA,TCA A79S XP_016857407.1
Gene
PKLR
Gene Name
pyruvate kinase, liver and RBC
There are no transcripts associated with this gene.

View Full Product Details