Product Details

SNP ID
rs200428997
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:153775761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCACACCTTTCTCATTCACGGCTC[C/G]CGGCGCTTTAGCTACTCAGAGGCGG
Phenotype
MIM: 611347 MIM: 604193
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
INTS3 PubMed Links

Gene Details

Gene
INTS3
Gene Name
integrator complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324475.1 470 Intron NP_001311404.1
NM_023015.4 470 Intron NP_075391.3
XM_005245459.2 470 Intron XP_005245516.1
XM_005245461.2 470 Intron XP_005245518.1
XM_006711490.3 470 Intron XP_006711553.1
XM_011509906.2 470 Intron XP_011508208.1
XM_017002132.1 470 Intron XP_016857621.1
Gene
SLC27A3
Gene Name
solute carrier family 27 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317929.1 470 Silent Mutation TCC,TCG S135S NP_001304858.1
NM_024330.1 470 Silent Mutation TCC,TCG S135S NP_077306.1

View Full Product Details