Product Details

SNP ID
rs200430087
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:47099198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCAGTGCTATCTCCTGGATCCT[C/T]TACTGCTTGGCAAAGTACCCTGAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP4Z1 PubMed Links

Gene Details

Gene
CYP4Z1
Gene Name
cytochrome P450 family 4 subfamily Z member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178134.2 Intron NP_835235.1

View Full Product Details