Product Details
- SNP ID
-
rs200443495
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:109280906 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACTCCGGCTGGTGAGTGGCATTC[A/G]GCTGGCAGGCCTGGGGATGGCCGAG
- Phenotype
-
MIM: 604265
MIM: 613126
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CELSR2
PubMed Links
Gene Details
- Gene
- CELSR2
- Gene Name
- cadherin EGF LAG seven-pass G-type receptor 2
There are no transcripts associated with this gene.
- Gene
- PSRC1
- Gene Name
- proline and serine rich coiled-coil 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005290.3 |
1004 |
Missense Mutation |
CCG,CTG |
P225L |
NP_001005290.1 |
NM_001032291.2 |
1004 |
Nonsense Mutation |
CGA,TGA |
R259* |
NP_001027462.1 |
NM_032636.7 |
1004 |
Nonsense Mutation |
CGA,TGA |
R259* |
NP_116025.1 |
XM_005271283.2 |
1004 |
Nonsense Mutation |
CGA,TGA |
R259* |
XP_005271340.1 |
XM_011542306.2 |
1004 |
Nonsense Mutation |
CGA,TGA |
R243* |
XP_011540608.1 |
XM_017002560.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R289* |
XP_016858049.1 |
XM_017002561.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R289* |
XP_016858050.1 |
XM_017002562.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R289* |
XP_016858051.1 |
XM_017002563.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R289* |
XP_016858052.1 |
XM_017002564.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R289* |
XP_016858053.1 |
XM_017002565.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R289* |
XP_016858054.1 |
XM_017002566.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R289* |
XP_016858055.1 |
XM_017002567.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R289* |
XP_016858056.1 |
XM_017002568.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R259* |
XP_016858057.1 |
XM_017002569.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R259* |
XP_016858058.1 |
XM_017002570.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R259* |
XP_016858059.1 |
XM_017002571.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R243* |
XP_016858060.1 |
XM_017002572.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R243* |
XP_016858061.1 |
XM_017002573.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R243* |
XP_016858062.1 |
XM_017002574.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R243* |
XP_016858063.1 |
XM_017002575.1 |
1004 |
Nonsense Mutation |
CGA,TGA |
R243* |
XP_016858064.1 |
XM_017002576.1 |
1004 |
Missense Mutation |
CCG,CTG |
P225L |
XP_016858065.1 |
XM_017002577.1 |
1004 |
Missense Mutation |
CCG,CTG |
P225L |
XP_016858066.1 |
View Full Product Details