Product Details

SNP ID
rs200508805
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:248239146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTGTCCCTTATGGACCTCATGC[C/T]CATCTGCACCACTCTACCCAAGATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2M4 PubMed Links

Gene Details

Gene
OR2M4
Gene Name
olfactory receptor family 2 subfamily M member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017504.1 218 Missense Mutation CCC,CTC P73L NP_059974.1

View Full Product Details