Product Details

SNP ID
rs200509081
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:37612821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCTGCTGCTGTTGCTGCCCCTT[G/T]CGTCTTCGAGAGCCAGCACCCGCCT
Phenotype
MIM: 609595
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RSPO1 PubMed Links

Gene Details

Gene
RSPO1
Gene Name
R-spondin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038633.3 1618 Silent Mutation CGA,CGC R242R NP_001033722.1
NM_001242908.1 1618 Silent Mutation CGA,CGC R242R NP_001229837.1
NM_001242909.1 1618 Silent Mutation CGA,CGC R215R NP_001229838.1
NM_001242910.1 1618 Silent Mutation CGA,CGC R179R NP_001229839.1
XM_006710583.3 1618 Silent Mutation CGA,CGC R242R XP_006710646.1

View Full Product Details