Product Details

SNP ID
rs200644020
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:248362100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCTCATGAACCATAGGGTGTGT[A/C]TCTTCCTGTCATCAGGCTGCTGGTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR2T4 PubMed Links

Gene Details

Gene
OR2T4
Gene Name
olfactory receptor family 2 subfamily T member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004696.1 Intron NP_001004696.1

View Full Product Details