Product Details

SNP ID
rs200671071
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:11649165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACTCAGCCAGCACGTTCTCGTG[A/C]TCGGACAGTAGCTTAACGGTGAGCT
Phenotype
MIM: 607112 MIM: 609111
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FBXO2 PubMed Links

Gene Details

Gene
FBXO2
Gene Name
F-box protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012168.5 1169 Missense Mutation GAG,GAT E226D NP_036300.2
Gene
FBXO44
Gene Name
F-box protein 44
There are no transcripts associated with this gene.

View Full Product Details