Product Details

SNP ID
rs202126222
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:92476182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCTGGCTGAATGCCTTGCCGCA[C/T]ACCTGGCACTTGTGAGGCTTCTCAC
Phenotype
MIM: 600871
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFI1 PubMed Links

Gene Details

Gene
GFI1
Gene Name
growth factor independent 1 transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127215.1 1484 Silent Mutation GTA,GTG V372V NP_001120687.1
NM_001127216.1 1484 Silent Mutation GTA,GTG V372V NP_001120688.1
NM_005263.3 1484 Silent Mutation GTA,GTG V372V NP_005254.2
XM_005270749.3 1484 Silent Mutation GTA,GTG V372V XP_005270806.1
XM_011541245.2 1484 Silent Mutation GTA,GTG V372V XP_011539547.1
XM_011541246.2 1484 Silent Mutation GTA,GTG V372V XP_011539548.1

View Full Product Details