Product Details

SNP ID
rs201916678
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:220102540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTTGAATATAATCCTGAGCATGT[C/T]AGTCGTTCAATATATGTAAAATTTC
Phenotype
MIM: 612801
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IARS2 PubMed Links

Gene Details

Gene
IARS2
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018060.3 899 Silent Mutation GTC,GTT V265V NP_060530.3

View Full Product Details