Product Details
- SNP ID
-
rs201881520
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:23310583 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGATGGGTTGGGAACCCCAGTTCTG[C/T]TGGTTGTTGGTCTGACGACGCTTGG
- Phenotype
-
MIM: 607201
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HNRNPR
PubMed Links
Gene Details
- Gene
- HNRNPR
- Gene Name
- heterogeneous nuclear ribonucleoprotein R
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001102397.2 |
1945 |
Intron |
|
|
NP_001095867.1 |
NM_001102398.2 |
1945 |
Silent Mutation |
CAA,CAG |
Q594Q |
NP_001095868.1 |
NM_001102399.2 |
1945 |
Intron |
|
|
NP_001095869.1 |
NM_001297620.1 |
1945 |
Silent Mutation |
CAA,CAG |
Q553Q |
NP_001284549.1 |
NM_001297621.1 |
1945 |
Intron |
|
|
NP_001284550.1 |
NM_001297622.1 |
1945 |
Intron |
|
|
NP_001284551.1 |
NM_005826.4 |
1945 |
Silent Mutation |
CAA,CAG |
Q591Q |
NP_005817.1 |
XM_005245711.4 |
1945 |
Silent Mutation |
CAA,CAG |
Q591Q |
XP_005245768.1 |
XM_011540471.2 |
1945 |
Silent Mutation |
CAA,CAG |
Q594Q |
XP_011538773.1 |
XM_011540472.2 |
1945 |
Intron |
|
|
XP_011538774.1 |
XM_011540474.2 |
1945 |
Silent Mutation |
CAA,CAG |
Q556Q |
XP_011538776.1 |
XM_011540475.2 |
1945 |
Silent Mutation |
CAA,CAG |
Q556Q |
XP_011538777.1 |
XM_011540476.2 |
1945 |
Silent Mutation |
CAA,CAG |
Q455Q |
XP_011538778.1 |
XM_011540477.2 |
1945 |
Silent Mutation |
CAA,CAG |
Q434Q |
XP_011538779.1 |
XM_017000008.1 |
1945 |
Silent Mutation |
CAA,CAG |
Q553Q |
XP_016855497.1 |
XM_017000009.1 |
1945 |
Silent Mutation |
CAA,CAG |
Q532Q |
XP_016855498.1 |
XM_017000010.1 |
1945 |
Silent Mutation |
CAA,CAG |
Q532Q |
XP_016855499.1 |
View Full Product Details