Product Details

SNP ID
rs202227038
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:161098821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACGGGCGTTGGCTCTGCGTGGTG[C/G]GCGGCTGGGACGGGTCTCGCCGCTT
Phenotype
MIM: 609607 MIM: 613466
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KLHDC9 PubMed Links

Gene Details

Gene
KLHDC9
Gene Name
kelch domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007255.2 431 Missense Mutation CGC,GGC R96G NP_001007256.1
NM_152366.4 431 Missense Mutation CGC,GGC R96G NP_689579.3
Gene
NECTIN4
Gene Name
nectin cell adhesion molecule 4
There are no transcripts associated with this gene.

Gene
PFDN2
Gene Name
prefoldin subunit 2
There are no transcripts associated with this gene.

View Full Product Details