Product Details

SNP ID
rs202079691
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:36393945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCAGCTTGACCTGATGCCCCCAA[A/C]GGTCCGTGTAGGTGACTTTGGCCAG
Phenotype
MIM: 609437
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LSM10 PubMed Links

Gene Details

Gene
LSM10
Gene Name
LSM10, U7 small nuclear RNA associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032881.2 402 Missense Mutation NP_116270.1
Gene
STK40
Gene Name
serine/threonine kinase 40
There are no transcripts associated with this gene.

View Full Product Details