Product Details

SNP ID
rs202228836
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:38998280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTGCTTCGGAAAGTCAACCTCA[C/T]TCCTCAGCACTCACAGCACCTAGCT
Phenotype
MIM: 615164
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKIRIN1 PubMed Links

Gene Details

Gene
AKIRIN1
Gene Name
akirin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136275.1 467 Silent Mutation CAC,CAT H110H NP_001129747.1
NM_024595.2 467 Silent Mutation CAC,CAT H110H NP_078871.1

View Full Product Details