Product Details

SNP ID
rs201924525
Assay Type
Functionally Tested
NCBI dbSNP Submissions
5
Location
Chr.1:71065109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAACTTGAACGGGAACCAGAAT[G/T]GGATGATCCAGATGATGACCTGTGG
Phenotype
MIM: 604347
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MIR186 PubMed Links

Gene Details

Gene
MIR186
Gene Name
microRNA 186
There are no transcripts associated with this gene.

Gene
ZRANB2
Gene Name
zinc finger RANBP2-type containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005455.4 1252 UTR 3 NP_005446.2
NM_203350.2 1252 Missense Mutation AAT,CAT N320H NP_976225.1
Gene
ZRANB2-AS1
Gene Name
ZRANB2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details