Product Details

SNP ID
rs201991921
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:10463097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATAAGCTCAGCTCTGGAGCCTG[C/T]TTCTCCCTCAGTGCAGTAAGGATGT
Phenotype
MIM: 601882
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DFFA PubMed Links

Gene Details

Gene
DFFA
Gene Name
DNA fragmentation factor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004401.2 842 Silent Mutation AAA,AAG K248K NP_004392.1
NM_213566.1 842 Silent Mutation AAA,AAG K248K NP_998731.1

View Full Product Details