Product Details

SNP ID
rs202110440
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:46402143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGCCCTGCCCCTGCCTCAGCTGG[C/T]GCAGAAGTTACACAGTAGAGAGCTG
Phenotype
MIM: 602935
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAAH PubMed Links

Gene Details

Gene
FAAH
Gene Name
fatty acid amide hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001441.2 330 Missense Mutation GCG,GTG A83V NP_001432.2

View Full Product Details