Product Details

SNP ID
rs202123513
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:225404421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTTACCACATGGGAGGGACCAC[A/G]CCAAGGCCATGATGAGATCACCCAA
Phenotype
MIM: 603341 MIM: 600024
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAH14 PubMed Links

Gene Details

Gene
DNAH14
Gene Name
dynein axonemal heavy chain 14
There are no transcripts associated with this gene.

Gene
LBR
Gene Name
lamin B receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002296.3 1674 Missense Mutation GCG,GTG A557V NP_002287.2
NM_194442.2 1674 Missense Mutation GCG,GTG A557V NP_919424.1
XM_005273125.2 1674 Missense Mutation GCG,GTG A515V XP_005273182.1
XM_011544185.2 1674 Missense Mutation GCG,GTG A557V XP_011542487.1

View Full Product Details