Product Details

SNP ID
rs201834450
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:236142980 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCGTCTTCTGGCTGCTCTACTGC[C/T]TCCCTGTGTGCCTGCAGTTTTTCAC
Phenotype
MIM: 604658
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR137B PubMed Links

Gene Details

Gene
GPR137B
Gene Name
G protein-coupled receptor 137B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003272.3 449 Missense Mutation CTC,TTC L120F NP_003263.1
XM_017002209.1 449 Missense Mutation CTC,TTC L120F XP_016857698.1
XM_017002210.1 449 Missense Mutation CTC,TTC L120F XP_016857699.1

View Full Product Details