Product Details

SNP ID
rs202098227
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:114774308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGAAGCAGTGTCCATTGAGTTT[C/T]CCTTTCTGGCTTGAAGAGACTCACT
Phenotype
MIM: 611656
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIKE1 PubMed Links

Gene Details

Gene
SIKE1
Gene Name
suppressor of IKBKE 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102396.1 667 Missense Mutation GAA,GGA E200G NP_001095866.1
NM_025073.2 667 Missense Mutation GAA,GGA E196G NP_079349.2

View Full Product Details