Product Details

SNP ID
rs201719480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45309612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTCTGAGGGGAGGCGTGCGCAG[A/G]TGCCTGCAGCAACAGTGTGAACAGA
Phenotype
MIM: 603897 MIM: 616104
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MATN4 PubMed Links

Gene Details

Gene
MATN4
Gene Name
matrilin 4
There are no transcripts associated with this gene.

Gene
RBPJL
Gene Name
recombination signal binding protein for immunoglobulin kappa J region like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281448.1 257 Silent Mutation AGA,AGG R59R NP_001268377.1
NM_001281449.1 257 Silent Mutation AGA,AGG R59R NP_001268378.1
NM_014276.3 257 Silent Mutation AGA,AGG R59R NP_055091.2
XM_011528521.2 257 Silent Mutation AGA,AGG R59R XP_011526823.1
XM_011528522.2 257 Silent Mutation AGA,AGG R59R XP_011526824.1
XM_011528523.2 257 Silent Mutation AGA,AGG R59R XP_011526825.1

View Full Product Details