Product Details

SNP ID
rs200514789
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:51004396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCATCTCGTTCTCGATGACGTA[A/G]AGCAGGGGCGCGAAGAGGGCGATGG
Phenotype
MIM: 603788
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KCNG1 PubMed Links

Gene Details

Gene
KCNG1
Gene Name
potassium voltage-gated channel modifier subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002237.3 1647 Silent Mutation NP_002228.2
XM_006723785.2 1647 Silent Mutation XP_006723848.1
XM_011528800.1 1647 Silent Mutation XP_011527102.1
XM_011528801.1 1647 Silent Mutation XP_011527103.1
XM_011528802.1 1647 Silent Mutation XP_011527104.1
XM_011528803.2 1647 Silent Mutation XP_011527105.1
XM_011528804.1 1647 Silent Mutation XP_011527106.1
XM_011528805.2 1647 Silent Mutation XP_011527107.1
XM_011528806.1 1647 Silent Mutation XP_011527108.1

View Full Product Details