Product Details

SNP ID
rs200666482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46174965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCGTAGGCTTCGGTGTCCTGCTC[A/G]CCGCCGCCTTCGTCGTTGTATTTGA
Phenotype
MIM: 609920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH22 PubMed Links

Gene Details

Gene
CDH22
Gene Name
cadherin 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021248.2 2630 Silent Mutation GGC,GGT G676G NP_067071.1
XM_011528994.2 2630 Silent Mutation GGC,GGT G676G XP_011527296.1
XM_011528995.1 2630 Silent Mutation GGC,GGT G555G XP_011527297.1

View Full Product Details