Product Details

SNP ID
rs202174178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2537277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCCTGTCTCTTACAGCACGAGGC[A/G]GAGTGAAAGGGCGGGTGAAGAGCGG
Phenotype
MIM: 606707
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMC2 PubMed Links

Gene Details

Gene
TMC2
Gene Name
transmembrane channel like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080751.2 58 Missense Mutation AGA,GGA R15G NP_542789.2
XM_005260660.3 58 Intron XP_005260717.1

View Full Product Details