Product Details

SNP ID
rs200735011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35707173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAACATTGTGTTGCAAGTGGCTG[A/C]ACAGAGGCAGCTGCAGCTAAAGCTG
Phenotype
MIM: 603485 MIM: 604739
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NFS1 PubMed Links

Gene Details

Gene
NFS1
Gene Name
NFS1 cysteine desulfurase
There are no transcripts associated with this gene.

Gene
RBM39
Gene Name
RNA binding motif protein 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242599.1 Intron NP_001229528.1
NM_001242600.1 Intron NP_001229529.1
NM_001323422.1 Intron NP_001310351.1
NM_001323423.1 Intron NP_001310352.1
NM_001323424.1 Intron NP_001310353.1
NM_004902.3 Intron NP_004893.1
NM_184234.2 Intron NP_909122.1
XM_006723890.3 Intron XP_006723953.1
XM_006723891.3 Intron XP_006723954.1
XM_011529111.2 Intron XP_011527413.1
XM_017028138.1 Intron XP_016883627.1
XM_017028139.1 Intron XP_016883628.1
XM_017028140.1 Intron XP_016883629.1
XM_017028141.1 Intron XP_016883630.1
XM_017028142.1 Intron XP_016883631.1
XM_017028143.1 Intron XP_016883632.1
XM_017028144.1 Intron XP_016883633.1
XM_017028145.1 Intron XP_016883634.1
XM_017028146.1 Intron XP_016883635.1
XM_017028147.1 Intron XP_016883636.1
Gene
ROMO1
Gene Name
reactive oxygen species modulator 1
There are no transcripts associated with this gene.

View Full Product Details