Product Details

SNP ID
rs200836567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18510918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGGAACGTGTGGCCTTCCAGCC[A/G]GCTGGAGGCTACAAGAATGGTTGTA
Phenotype
MIM: 610512
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC23B PubMed Links

Gene Details

Gene
SEC23B
Gene Name
Sec23 homolog B, coat complex II component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172745.1 544 Missense Mutation CAG,CGG Q28R NP_001166216.1
NM_001172746.1 544 Missense Mutation CAG,CGG Q28R NP_001166217.1
NM_006363.4 544 Missense Mutation CAG,CGG Q28R NP_006354.2
NM_032985.4 544 Missense Mutation CAG,CGG Q28R NP_116780.1
NM_032986.3 544 Missense Mutation CAG,CGG Q28R NP_116781.1
XM_017027593.1 544 Missense Mutation CAG,CGG Q28R XP_016883082.1

View Full Product Details