Product Details

SNP ID
rs200588273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2794191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAGGGCTCTTAAACCGCAGGTAT[C/G]AATCCTTCTGTCCCCTTAGCCGCTC
Phenotype
MIM: 609555
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CPXM1 PubMed Links

Gene Details

Gene
CPXM1
Gene Name
carboxypeptidase X (M14 family), member 1
There are no transcripts associated with this gene.

View Full Product Details