Product Details

SNP ID
rs200555822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62216042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCTCCAGGGAGCTGTGGGGCTG[A/G]ATTTTGAGCTTCTGGGGGCAGAGCA
Phenotype
MIM: 604525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HRH3 PubMed Links

Gene Details

Gene
HRH3
Gene Name
histamine receptor H3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007232.2 1302 Silent Mutation ATC,ATT I434I NP_009163.2
XM_005260266.3 1302 Silent Mutation ATC,ATT I434I XP_005260323.1
XM_017027623.1 1302 Silent Mutation ATC,ATT I380I XP_016883112.1

View Full Product Details