Product Details
- SNP ID
-
rs201764439
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:17494338 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGTATAGATGGTTCCTCTGCACC[A/G]GGTGTGACCATGGCACAGGGTCTCC
- Phenotype
-
MIM: 603307
MIM: 162151
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BFSP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6080718] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BFSP1
- Gene Name
- beaded filament structural protein 1
- Gene
- PCSK2
- Gene Name
- proprotein convertase subtilisin/kexin type 2
There are no transcripts associated with this gene.
View Full Product Details