Product Details

SNP ID
rs199596476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58153614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAACTGGGGGTTTTTAACAACC[A/C]CTTTTGAGGAGGTAAATATAAATTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C20orf85 PubMed Links

Gene Details

Gene
C20orf85
Gene Name
chromosome 20 open reading frame 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178456.2 177 Missense Mutation ACT,CCT T47P NP_848551.1

View Full Product Details