Product Details

SNP ID
rs202184327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29008002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCTCTGCTGACATAGCCAGAGG[C/T]GTGTTCTCTAACCCACTCTGTGGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RWDD2B PubMed Links

Gene Details

Gene
RWDD2B
Gene Name
RWD domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320724.1 676 Missense Mutation ACC,GCC T133A NP_001307653.1
NM_016940.2 676 Missense Mutation ACC,GCC T162A NP_058636.1

View Full Product Details