Product Details

SNP ID
rs199655078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32737257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCATTAATTACAAAATTACCTT[A/G]CATTTCTTTTTTCCACATCAGAACA
Phenotype
MIM: 604297
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAXBP1 PubMed Links

Gene Details

Gene
PAXBP1
Gene Name
PAX3 and PAX7 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013329.3 2823 Intron NP_037461.2
NM_016631.3 2823 Missense Mutation GCA,GTA A878V NP_057715.2
XM_006724066.1 2823 Missense Mutation GCA,GTA A843V XP_006724129.1
XM_011529804.2 2823 Intron XP_011528106.1
Gene
PAXBP1-AS1
Gene Name
PAXBP1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SYNJ1
Gene Name
synaptojanin 1
There are no transcripts associated with this gene.

View Full Product Details