Product Details

SNP ID
rs199509733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44601709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGCCCGGCCTGCTGTGGCCCCA[C/T]CTCAACCCAGAAGTCCAGCTGCTGA
Phenotype
MIM: 612920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTAP10-6 PubMed Links

Gene Details

Gene
KRTAP10-6
Gene Name
keratin associated protein 10-6
There are no transcripts associated with this gene.

Gene
KRTAP10-7
Gene Name
keratin associated protein 10-7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198689.2 1113 Missense Mutation ACC,ATC T363I NP_941962.1
Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 1113 Intron NP_001258966.1
NM_144991.2 1113 Intron NP_659428.2

View Full Product Details