Product Details

SNP ID
rs199512349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37625278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGTTGGTTGAGTTTGCTGGATA[A/C]AGACCAACTCAGGGGCAGCTCTGCC
Phenotype
MIM: 600877
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNJ6 PubMed Links

Gene Details

Gene
KCNJ6
Gene Name
potassium voltage-gated channel subfamily J member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002240.4 1735 Missense Mutation NP_002231.1

View Full Product Details