Product Details
- SNP ID
-
rs200242899
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:42315361 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGACCAGCCCCAGCATGCAGAGC[A/G]CTCTGGCAGCCATGACCACCGTGGG
- Phenotype
-
MIM: 600633
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TFF3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11701143] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TFF3
- Gene Name
- trefoil factor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003226.3 |
236 |
Missense Mutation |
|
|
NP_003217.3 |
View Full Product Details