Product Details
- SNP ID
-
rs201050988
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:42403995 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACGCCAAGGTCTCCAACAAGCTCA[A/G]GAGCCGCAGCAGCCCCTCGCTCCTG
- Phenotype
-
MIM: 605511
MIM: 605736
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMPRSS3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2277798] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMPRSS3
- Gene Name
- transmembrane protease, serine 3
There are no transcripts associated with this gene.
- Gene
- UBASH3A
- Gene Name
- ubiquitin associated and SH3 domain containing A
View Full Product Details