Product Details

SNP ID
rs201420116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:30541663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGAATCAGCAAGTTTTTTAGTA[A/G]AATCCAGAGAATCCATATCCTTCAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP19-6 PubMed Links

Gene Details

Gene
KRTAP19-6
Gene Name
keratin associated protein 19-6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303120.1 202 Intron NP_001290049.1
NM_181612.3 202 Silent Mutation TTC,TTT F57F NP_853643.1

View Full Product Details