Product Details

SNP ID
rs200227163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41578857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAACTCTGTTTTCAGGGCTTCCA[C/T]GAACTTCTCCCGGATCTTCTCTTTC
Phenotype
MIM: 601786
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSDC2 PubMed Links

Gene Details

Gene
CSDC2
Gene Name
cold shock domain containing C2
There are no transcripts associated with this gene.

Gene
PMM1
Gene Name
phosphomannomutase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002676.2 432 Missense Mutation ATG,GTG M167V NP_002667.2
XM_005261638.4 432 Missense Mutation ATG,GTG M70V XP_005261695.1
XM_011530229.1 432 Missense Mutation ATG,GTG M210V XP_011528531.1
XM_011530230.2 432 Missense Mutation ATG,GTG M172V XP_011528532.1
XM_011530231.2 432 Missense Mutation ATG,GTG M97V XP_011528533.1
XM_011530232.1 432 Missense Mutation ATG,GTG M70V XP_011528534.1

View Full Product Details