Product Details
- SNP ID
-
rs200614375
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:30263945 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTTCCTGGAGGGTCTGGTCCTGC[A/G]CACCCCCTTCCTCAGGGCCTGGTGG
- Phenotype
-
MIM: 165095
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OSM
PubMed Links
Gene Details
- Gene
- OSM
- Gene Name
- oncostatin M
View Full Product Details