Product Details

SNP ID
rs200749842
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46274000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCTGTGAAGACAATGGTGGAGA[C/T]TTCAAGAACCCAGCCGCTGACAAGG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TTC38 PubMed Links

Gene Details

Gene
TTC38
Gene Name
tetratricopeptide repeat domain 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017931.2 372 Missense Mutation ACT,ATT T99I NP_060401.2
XM_011530259.1 372 Missense Mutation ACT,ATT T34I XP_011528561.1
XM_011530260.2 372 Missense Mutation ACT,ATT T99I XP_011528562.1

View Full Product Details