Product Details
- SNP ID
-
rs201085386
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:50625188 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAATGGCAGCAAGCTGGGCGGGGG[A/G]TGCAGCCAGGATGACAGCAGATCTG
- Phenotype
-
MIM: 607574
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARSA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6151428] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARSA
- Gene Name
- arylsulfatase A
View Full Product Details