Product Details

SNP ID
rs201693608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50625378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTCTCACCAGGGTCCTTGGACA[C/G]GTCATAGAGCAGCGGGGGCTCATGA
Phenotype
MIM: 607574
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARSA PubMed Links

Gene Details

Gene
ARSA
Gene Name
arylsulfatase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000487.5 1566 Missense Mutation CTG,GTG L433V NP_000478.3
NM_001085425.2 1566 Missense Mutation CTG,GTG L433V NP_001078894.2
NM_001085426.2 1566 Missense Mutation CTG,GTG L433V NP_001078895.2
NM_001085427.2 1566 Missense Mutation CTG,GTG L433V NP_001078896.2
NM_001085428.2 1566 Missense Mutation CTG,GTG L347V NP_001078897.1
XM_011530690.2 1566 Missense Mutation CTG,GTG L347V XP_011528992.1
XM_011530691.2 1566 UTR 3 XP_011528993.1
XM_017028800.1 1566 Missense Mutation CTG,GTG L471V XP_016884289.1

View Full Product Details