Product Details

SNP ID
rs199847760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50580390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTCATTGTGGCAGAAGACGACT[C/G]GCGATGGGGTAGACTCTAGTAACTT
Phenotype
MIM: 612395 MIM: 601987
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHKB PubMed Links

Gene Details

Gene
CHKB
Gene Name
choline kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005198.4 922 Missense Mutation CCA,CGA P235R NP_005189.2
Gene
CHKB-AS1
Gene Name
CHKB antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
CHKB-CPT1B
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
CPT1B
Gene Name
carnitine palmitoyltransferase 1B
There are no transcripts associated with this gene.

View Full Product Details