Product Details

SNP ID
rs199718994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20116227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGCCAGGCGCACACTGAGTGGG[C/T]GGCCTTTCCAGAGGGCACCATGCAA
Phenotype
MIM: 609030 MIM: 601180 MIM: 611151
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGCR8 PubMed Links

Gene Details

Gene
DGCR8
Gene Name
DGCR8 microprocessor complex subunit
There are no transcripts associated with this gene.

Gene
MIR6816
Gene Name
microRNA 6816
There are no transcripts associated with this gene.

Gene
RANBP1
Gene Name
RAN binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278639.1 606 Missense Mutation CGG,TGG R15W NP_001265568.1
NM_001278640.1 606 Intron NP_001265569.1
NM_001278641.1 606 Intron NP_001265570.1
NM_002882.3 606 Intron NP_002873.1
XM_011530289.2 606 Intron XP_011528591.1
XM_011530290.2 606 Intron XP_011528592.1
XM_011530291.2 606 Intron XP_011528593.1
XM_017028890.1 606 Intron XP_016884379.1
XM_017028891.1 606 Intron XP_016884380.1
XM_017028892.1 606 Intron XP_016884381.1
Gene
TRMT2A
Gene Name
tRNA methyltransferase 2 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257994.1 606 Missense Mutation CAC,CGC H137R NP_001244923.1
NM_022727.5 606 Missense Mutation CAC,CGC H137R NP_073564.3
NM_182984.4 606 Missense Mutation CAC,CGC H137R NP_892029.2
XM_011530139.2 606 Missense Mutation CAC,CGC H137R XP_011528441.1
XM_011530141.1 606 Intron XP_011528443.1
XM_011530142.2 606 Intron XP_011528444.1
XM_017028770.1 606 Missense Mutation CAC,CGC H137R XP_016884259.1

View Full Product Details