Product Details

SNP ID
rs201229776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29260190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACGGCCTGTGGCTGCCAGTGCC[A/C]CCACCGCCTGCTCCGTAGGGAAGCC
Phenotype
MIM: 608926 MIM: 133450
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EMID1 PubMed Links

Gene Details

Gene
EMID1
Gene Name
EMI domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267895.1 1333 Intron NP_001254824.1
NM_133455.3 1333 Intron NP_597712.2
XM_005261328.2 1333 Intron XP_005261385.1
XM_005261329.2 1333 Intron XP_005261386.1
XM_011529868.2 1333 Intron XP_011528170.1
XM_011529869.2 1333 Intron XP_011528171.1
XM_011529870.2 1333 Intron XP_011528172.1
XM_011529871.2 1333 Intron XP_011528173.1
XM_011529872.2 1333 Intron XP_011528174.1
XM_011529873.2 1333 Intron XP_011528175.1
XM_011529874.2 1333 Intron XP_011528176.1
XM_011529875.2 1333 Intron XP_011528177.1
XM_011529876.1 1333 Intron XP_011528178.1
XM_017028589.1 1333 Intron XP_016884078.1
Gene
EWSR1
Gene Name
EWS RNA binding protein 1
There are no transcripts associated with this gene.

Gene
RHBDD3
Gene Name
rhomboid domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006724224.3 1333 Missense Mutation GGG,GTG G216V XP_006724287.1
XM_011530107.2 1333 Missense Mutation GGG,GTG G344V XP_011528409.1
XM_017028749.1 1333 Missense Mutation GGG,GTG G347V XP_016884238.1
XM_017028750.1 1333 Missense Mutation GGG,GTG G211V XP_016884239.1

View Full Product Details